Product Image
Thermo Scientific™
Remel™ Enhanced Gram Safranin
Thermo Scientific™ enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R402375/Pk.
R40238Each
Catalog number R40237
Price (USD)
-
Quantity:
5/Pk.
Thermo Scientific™ enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
Specifications
Quantity5/Pk.
TypeSafranin
Volume (Metric)5 x 250 mL
Unit SizeEach