Product Image
Thermo Scientific™
CN Selective Supplement (Cefsulodin, Novobiocin)
Selectively isolate Yersinia enterocolitica using CN Selective Supplement (Cefsulodin, Novobiocin).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormQuantity
R450031Freeze-dried10 mL
R45050Liquid1 mL
Catalog number R450031
Price (USD)
-
Form:
Freeze-dried
Quantity:
10 mL
Selectively isolate Yersinia enterocolitica using Thermo Scientific™ Remel™ CN Selective Supplement (Cefsulodin, Novobiocin). This supplement is used in the preparation of CIN agar base which makes it selective and a differential medium for isolation of Y. enterocolitica. Supplementation with CN supplement improves inhibition of normal enteric organisms such as Escherichia coli, Klebsiella pneumoniae, Proteus mirabilis and Pseudomonas aeruginosa, as well as gram-positive organisms.
Selectively isolate Yersinia enterocolitica using CN Selective Supplement (Cefsulodin, Novobiocin).
The complete medium of CIN Agar formulation of Schiemann contains this highly selective supplement which is recommended for use in the isolation and cultivation of Yersinia enterocolitica from clinical specimens and food.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Quantity10 mL
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach