Product Image
Thermo Scientific™
Remel™ Buffered CYE Supplement
The Remel Buffered CYE Supplement (L-Cysteine HCl) is lyophilized.
Have Questions?
Catalog NumberFor Use With (Application)
R45005Used with CYE Agar Pour Tube
Catalog number R45005
Price (USD)
-
For Use With (Application):
Used with CYE Agar Pour Tube
Remel Buffered CYE Supplement (L-Cysteine HCl) is lyophilized.
  • Part No. R45005 is for use with Buffered CYE Agar Pour Tube, Part No. R09300. Part No. R450041 is for use with Buffered CYE Agar Base, Part No. R452352
For Laboratory Use Only
Specifications
For Use With (Application)Used with CYE Agar Pour Tube
Quantity10 x 1 mL
FormFreeze-dried
Product TypeMicrobiology Media
Unit SizeEach