Product Image
Thermo Scientific™
Remel™ Vitamin K, Hemin Solution
Remel Vitamin K, Hemin Solution is used in anaerobic media.
Have Questions?
Catalog NumberQuantity
R45095110 mL
Catalog number R450951
Price (USD)
-
Quantity:
10 mL
Remel Vitamin K, Hemin Solution is used in anaerobic media.
Specifications
ConcentrationVitamin K: 0.5mg/10mL. Hemin: 5mg/10mL.
DescriptionVitamin K, Hemin Solution
Quantity10 mL
FormLiquid
Product TypeSolution
Unit SizeEach