Product Image
Thermo Scientific™
Remel™ Buffered Peptone Water
Thermo Scientific™ Remel Buffered Peptone Water is used for the non-selective pre-enrichment of Salmonella spp. from foods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R45267610 kg
R452672500 g
R4526742.5 kg
Catalog number R452676
Price (USD)
-
Quantity:
10 kg
Thermo Scientific™ Remel Buffered Peptone Water is used for the non-selective pre-enrichment of Salmonella spp. from foods.
For Laboratory Use Only
Specifications
DescriptionBuffered Peptone Water
Quantity10 kg
FormPowder
Product TypePeptone Water
Unit SizeEach