Product Image
Thermo Scientific™
CLED Agar (Dehydrated)
Differentiate and enumerate microorgansims in urine specimens using CLED Agar.
Have Questions?
Catalog NumberQuantity
R452882500 g
Catalog number R452882
Price (USD)
-
Quantity:
500 g
Differentiate and enumerate microorgansims in urine specimens using Thermo Scientific™ Remel™ CLED Agar (Dehydrated). Mackey and Sandys modified the solid culture medium for the use of with urine cultures first developed in 19601 by substituting lactose and sucrose for mannitol and increasing the concentration of brom thymol blue and agar2. Further addition of cystine and deletion of sucrose resulted in Cystine Lactose Electrolyte Deficient (CLED) Agar which detects bacteriuria by quantitative culture of urine3.
Differentiate and enumerate microorgansims in urine specimens using CLED Agar.
  • Ease of Differentiation: Use of bromthymol blue as a pH indicator helps in differentiating lactose fermenters from non-fermenters.
  • Recommended for quantitative measurements by inoculating the surface of the medium with appropriate dilutions of the urine sample.
This medium is recommended for use in qualitative procedures for the cultivation of microorganisms from urine specimens. It contains beef extract which supplies carbohydrates and vitamins, casein peptone which supplies amino acids, nitrogenous compounds and peptides necessary for the growth of bacteria. Cystine enhances the growth of cystine-dependant coliforms. Brom thymol blue is a pH indicator which differentiates lactose fermenters producing yellow colonies from lactose nonfermenters. Electrolytes are reduced in order to restrict the swarming of Proteus.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
  1. Sandys, G.H. 1960. J. Med. Lab. Technol. 17:224-233.
  2. Mackey, J.P. and G.H. Sandys. 1965. Br. Med. J. 2:1286-1288.
  3. Mackey, J.P. and G.H. Sandys. 1966. Br. Med. J. 1:1173.
Specifications
DescriptionCLED Agar
Quantity500 g
FormPowder
Product TypeAgar
Unit SizeEach