Product Image
Thermo Scientific™
Remel™ Modified EMB Agar
The Remel Modified EMB Agar (HHT) is used for selective and differential isolation of gram-negative bacilli.
Have Questions?
Catalog NumberQuantity
R453392500 g
Catalog number R453392
Price (USD)
-
Quantity:
500 g
Remel Modified EMB Agar (HHT) is used for selective and differential isolation of gram-negative bacilli.
For Laboratory Use Only
Specifications
DescriptionEMB Agar, Modified
Quantity500 g
FormPowder
Product TypeAgar
Unit SizeEach