Remel™ Levine EMB Agar, Dehydrated
Thermo Scientific™
Remel™ Levine EMB Agar, Dehydrated
Remel EMB Agar, Levine (Dehydrated) provides selective isolation and differentiation of gram-negative enteric bacilli from clinical and industrial samples.
Have Questions?
Catalog NumberQuantity
R453402500 g
Catalog number R453402
Price (USD)
-
Quantity:
500 g
Remel EMB Agar, Levine (Dehydrated) provides selective isolation and differentiation of gram-negative enteric bacilli from clinical and industrial samples.
Specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF, IDF, USP.
DescriptionEMB Agar, Levine
Quantity500 g
FormPowder
Product TypeAgar
Unit SizeEach