Product Image
Thermo Scientific™
Remel™ EMB Agar Base
Remel EMB Agar Base is used for selective and differential isolation of gram-negative bacilli.
Have Questions?
Catalog NumberQuantity
R453412100 g
Catalog number R453412
Price (USD)
-
Quantity:
100 g
Remel EMB Agar Base is used for selective and differential isolation of gram-negative bacilli.
For Laboratory Use Only
Specifications
DescriptionEugon Agar; 100 g
Quantity100 g
FormPowder
Product TypeAgar
Unit SizeEach