Fluid Thioglycollate Medium (Dehydrated)
Actual product may vary
Thermo Scientific™
Fluid Thioglycollate Medium (Dehydrated)
Cultivate aerobes and anaerobes and carry out sterility testing using Fluid Thioglycollate Medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R45345610 kg
R453452500 g
Catalog number R453456
Price (USD)
-
Quantity:
10 kg
Cultivate aerobes and anaerobes and carry out sterility testing using Thermo Scientific™ Remel™ Fluid Thioglycollate Medium (Dehydrated). This general-purpose medium was first described by Brewer in 19401 consisting of a small amount of agar and a reducing substance to initiate the growth of anaerobes2 and for sterility testing. This medium is recommended by the United States Pharmacopeia (USP) and the AOAC International (AOAC), the American Public Health Association (APHA) and Food and Drug Administration (FDA) in the Bacteriological Analytical Manual Online3-6.
Cultivate aerobes and anaerobes using Fluid Thioglycollate Medium.
  • Recommended by APHA, AOAC, USP and FDA.
  • Ease of Identification: Resazurin is an oxidation-reduction indicator which turns pink when oxidation is increased.
This medium is used for qualitative procedures. This medium contains casein peptone and cystine that provides nitrogen and carbon compounds, agar that impedes oxygen diffusion, and sodium thioglycollate, which is a reducing agent that maintains a low oxygen tension in anaerobic medium. Yeast extract supplies B-complex vitamins and enhances the growth of microorganisms.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
  1. Brewer, J.H. 1940. JAMA. 115:598-600.
  2. Macfaddin, J.F. 1985. Media for Isolation-Cultivation-Identification-Maintenance of Medical Bacteria. Vol. 1. Williams and Wilkins, Baltimore, MD.
  3. The United States Pharmacopeia. 2009. 32nd United States Pharmacopeial Convention, Rockville, MD.
  4. Horwitz, W. 2002. Official Methods of Analysis of AOAC International. 17th, 1st Rev. AOAC International, Washington, D.C.
  5. Food and Drug Administration. 2000. Bacteriological Analytical Manual Online. AOAC International, Gaithersburg, MD.
  6. Downes, F.P. and K. Ito. 2001. Compendium of Methods for the Microbiological Examination of Foods. 4th American Public Health Association, Washington, D.C.
Specifications
DescriptionFluid Thioglycollate Medium
Quantity10 kg
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach