Product Image
Thermo Scientific™
Remel™ Lauryl Tryptose Broth with MUG
The Remel Lauryl Tryptose Broth with MUG is used in fluorogenic assays for detection of Escherichia coli in water, wastewater and foods.
Have Questions?
Catalog number R453672
Price (USD)
-
Remel Lauryl Tryptose Broth with MUG is used in fluorogenic assays for the detection of Escherichia coli in water, wastewater and foods.
Specifications
DescriptionLauryl Tryptose Broth w/MUG
FormatPowder
Quantity500 g
Product TypeDehydrated Culture Media
Unit SizeEach