Product Image
Thermo Scientific™
Letheen Broth Modified (Dehydrated)
Effectively carry out microbiological testing of cosmetics using Letheen Broth Modified.
Have Questions?
Catalog NumberQuantity
R453732500 g
Catalog number R453732
Price (USD)
-
Quantity:
500 g
Effectively carry out microbiological testing of cosmetics using Thermo Scientific™ Remel™ Letheen Broth Modified (Dehydrated). Weber and Black modified Tryptose Glucose Extract Broth by incorporating lecithin and polysorbate 80 as a result of their study on quaternary ammonium compounds1,2. Letheen Broth Modified is based on the formula in the Bacteriological Analytical Manual for use in the microbiological testing of cosmetics3. It is recommended by the Association of Official Analytical Chemists (AOAC) for cultivation of microorganisms from cosmetic samples4.
Effectively carry out microbiological testing of cosmetics using Letheen Broth Modified.
  • Recommended by AOAC for cultivation of microorganisms from cosmetic samples4. 
  • Based on the formula in the Bacteriological Analytical Manual for use in the microbiological testing of cosmetics3.
This medium contains meat peptone and beef extract which supply nutrition necessary for the growth of bacteria. Letheen broth modified contains additional casein peptone which supplies nitrogen for enhanced growth of microorganisms. Yeast extract provides B-complex vitamins. Sodium bisulfite neutralizes quaternary ammonium compounds, lecithin and polysorbate 80 partially inactivate quaternary ammonium compounds, phenolic compounds, formalin, hexachlorophene and ethanol.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
  1. Weber, G.R. and L.A. Black. 1948. Soap Sanitary Chem. 24:134-137, 151, 153, 155.
  2. Weber, G.R. and L.A. Black. 1948. Am. J. Public Health. 38:1405-1417.
  3. Food and Drug Administration. 2001. Bacteriological Analytical Manual Online. Chapter 23. Revised August 2001. Authors: A.D. Hitchins, T.T. Tran, and J.E. McCarron. AOAC International, Gaithersburg, MD.
  4. Horwitz, W. 2002. Official Methods of Analysis of AOAC Internantional. 17th ed., 1st rev. AOAC International, Washington, D.C.
Specifications
DescriptionLetheen Broth Modified
Quantity500 g
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach