Product Image
Thermo Scientific™
Remel™ Lysine Iron Agar
Differentiate microorganisms based on lysine decarboxylase and hydrogen sulfide production using Lysine Iron Agar (Dehydrated).
Have Questions?
Catalog NumberQuantity
R453772500 g
Catalog number R453772
Price (USD)
-
Quantity:
500 g
Differentiate microorganisms based on lysine decarboxylase and hydrogen sulfide production using Thermo Scientific™ Remel™ Lysine Iron Agar (Dehydrated). In 1961, Lysine Iron Agar (Dehydrated) was developed to detect lactose positive Arizona strains implicated in outbreaks of food-borne disease1. In 1966, Johnson et al. recommended LIA to help identify Salmonella spp2. Later, Ewing recommended the use of LIA with TSI to detect enteric pathogens in routine examination of stools3.
Lysine as a substrate is used for detection of lysine decarboxylase and lysine deaminase. When Salmonella spp. decarboxylates lysine, the amine is converted to cadaverine resulting in a purple butt (alkaline). When Proteus spp. deaminates lysine, the amine is converted to α-ketocarboxylic acid resulting in the slant turning red.
Interpretation of test:
  1. Lysine Decarboxylation (detected in butt)
    • Positive test – Purple slant/purple butt (alkaline), the butt reaction may be masked by H2S production
    • Negative test – Purple slant/yellow butt (acid), fermentation of glucose only
  2. Lysine Deamination (detected on slant)
    • Positive test – Red slant
    • Negative test – Slant remains purple
  3. H2S Production
    • Positive test – Black precipitate
    • Negative test – No black color development
The medium contains gelatin peptone and yeast extract that provide nitrogen, amino acids, and vitamins necessary for bacterial growth. Dextrose is the fermentable carbohydrate source and brom cresol purple is a pH indicator. Sodium thiosulfate and ferric ammonium citrate serve as indicators when H2S is produced, resulting in a black precipitate in the butt of the tube.
For Laboratory Use Only
  1. Edwards, P.R. and M.A. Fife. 1961. Appl. Microbiol. 9:478.
  2. Johnson, J.G., L.J. Kunz, W. Barron, and W.H. Ewing. 1966. Appl. Microbiol. 14:212-217.
  3. Ewing, W.H. 1986. Identification of Enterobacteriaceae. 4th ed. Elsevier, New York, NY.
Specifications
DescriptionLysine Iron Agar
Quantity500 g
FormPowder
Product TypeAgar
Unit SizeEach