Product Image
Thermo Scientific™
M Broth (Dehydrated)
Detect and cultivate of Salmonella in dried foods and feeds using M Broth.
Have Questions?
카탈로그 번호수량
R453782500 g
카탈로그 번호 R453782
제품 가격(KRW)
-
수량:
500 g
Detect and cultivate Salmonella in dried foods and feeds using Thermo Scientific™ Remel™ M Broth (Dehydrated). This enrichment medium was first formulated in 1969 by Sperber and Deibel using an accelerated serology technique1. This media increases rapid growth of Salmonella, improve flagella development and prevents nonspecific agglutination of bacteria cells used in serology techniques. It is currently recommended by the AOAC International Official Methods of Analysis, and Microbiology Laboratory Guidebook for the detection of Salmonella in foods2-4.
Accelerated detection and cultivation of Salmonella in dried foods and feeds using M Broth.
  • Accelerated Serology Technique: For detection of Salmonella in dried foods and feeds which prevents delayed transportation of bulk feeds to the market place unlike previous methods.
  • Recommended by the AOAC International Official Methods of Analysis, and Microbiology Laboratory Guidebook for the detection of Salmonella in foods2-4.
This medium is used for qualitative procedures. This medium contains casein peptone that provides amino acids and nitrogenous compounds necessary for the bacterial growth and yeast extract that provides B-complex vitamins. Mannose is the carbon source and serves to prevent fimbrial agglutination in serological techniques. Polysorbate 80 acts as a surfactant and dispersing agent.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
  1. Sperber, W.H. and R.H. Deibel. 1969. Appl. Microbiol. 17:533-539.
  2. Downes, F.P. and K. Ito. 2001. Compendium of Methods for the Microbiological Examination of Foods. 4th American Public Health Association, Washington, D.C.
  3. AOAC International. 2002. Official Methods of Analysis of AOAC International. 17th, 1st rev. AOAC International, Washington, D.C.
  4. United States Department of Agriculture. 2008. Laboratory Guidebook.
사양
설명M Broth
수량500 g
형태Powder
제품 유형Dehydrated Culture Media
Unit SizeEach