Remel™ MacConkey Agar
Actual product may vary
Thermo Scientific™
Remel™ MacConkey Agar
The Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R45380610 kg
R453802500 g
R4538042.5 kg
Catalog number R453806
Price (USD)
-
Quantity:
10 kg
Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Specifications
DescriptionMacConkey Agar
Media TypeMacConkey Agar
Quantity10 kg
FormPowder
Product TypeNutrient Agar
Unit SizeEach