Product Image
Thermo Scientific™
Mueller Hinton Agar
The Thermo Scientific™ Remel Mueller Hinton Agar is used for antimicrobial disk diffusion susceptibility testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R454082500 g
R4540842.5 kg
R45408610 kg
Catalog number R454082
Price (MXN)
-
Quantity:
500 g
The Thermo Scientific™ Remel Mueller Hinton Agar is used for antimicrobial disk diffusion susceptibility testing.
Specifications
Certifications/ComplianceConforms to CLSI standards. Industrial Reference: BAM, COMPF.
DescriptionMueller Hinton Agar
Media TypeMueller Hinton Agar
Quantity500 g
FormPowder
Product TypeNutrient Agar
Unit SizeEach