Product Image
Thermo Scientific™
Remel™ Sabouraud Dextrose Agar, Emmons
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons is used for cultivation and isolation of yeast and fungi.
Have Questions?
Catalog NumberQuantity
R454472Each
Catalog number R454472
Price (USD)
-
Quantity:
Each
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons is used for cultivation and isolation of yeast and fungi.
R454472 is not intended for IVD use.
Specifications
DescriptionSabouraud Dextrose Agar, Emmons
QuantityEach
FormPowder
Product TypeAgar
Unit SizeEach