Product Image
Thermo Scientific™
Remel™ Tetrathionate Broth Base
Remel Tetrathionate Broth Base is selective enrichment medium for use with iodine for the recovery of Salmonella species.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R4548242.5 kg
R454822500 g
Catalog number R454824
Price (USD)
-
Quantity:
2.5 kg
Remel Tetrathionate Broth Base is selective enrichment medium for use with iodine for the recovery of Salmonella species.
Also available: Brilliant Green for Tetrathionate, Part No. R114080 and Iodine For Tetrathionate, Part No. R114350.
For Laboratory Use Only
Specifications
DescriptionTetrathionate Broth Base
Quantity2.5 kg
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach