Product Image
Thermo Scientific™
Remel™ Eugonic Broth with Lecithin, Polysorbate 80, Triton X-100™, 500g
Remel Eugonic Broth with Lecithin, Polysorbate 80, Triton† is used for cultivation of fastidious microorganisms.
Have Questions?
Catalog NumberQuantity
R458012500 g
Catalog number R458012
Price (USD)
-
Quantity:
500 g
Remel Eugonic Broth with Lecithin, Polysorbate 80, Triton† is used for cultivation of fastidious microorganisms.
  • Triton X-100 is a surfactant which helps reduce adhesive forces that may be present in samples
For Laboratory Use Only
Specifications
DescriptionEugonic Broth w/Lecithin, Polysorbate 80; Triton X-100
Quantity500 g
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach