Product Image
Thermo Scientific™
Remel™ XLT4 Agar Base
Remel XLT4 Agar Base is used with Tergitol 4 for the isolation of non-typhi Salmonella species.
Have Questions?
Catalog NumberQuantity
R459802500 g
Catalog number R459802
Price (USD)
-
Quantity:
500 g
Remel XLT4 Agar Base is used with Tergitol 4 for the isolation of non-typhi Salmonella species.
Also available: Tergitol 4, Part No. R450880.
For Laboratory Use Only
Specifications
DescriptionXLT4 Agar Base
Quantity500 g
FormPowder
Product TypeAgar
Unit SizeEach