This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Candida tropicalis ATCC™ 750™
Thermo Scientific™
Culti-Loops™ Candida tropicalis ATCC™ 750™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46012405 Loops/Pack
Catalog number R4601240
Price (USD)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®750TM*
DescriptionCandida tropicalis
FormatCulti-Loops
Quantity5 Loops/Pack
Strain DesignationsWDCM00112, 1909, ATCC 7349, ATCC4563,CBS 94, CCRC 20520, IFO 1070, IFO 1400, JCM 1541, NRRL Y-12699, NRRL Y-12968, NRRL Y-607]
Product TypeCulti-Loops Quality Control Organism
SpeciesCandida species
Target Organism ClassFungi
Test TypeQualitative QC
Unit SizeEach