This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops&trade; <i>Bacteroides fragilis</i> ATCC&reg; 25285&trade;
Thermo Scientific™

Culti-Loops™ Bacteroides fragilis ATCC® 25285™

Perform quality control testing and performance evaluation of microbiological procedures with these ready-to-use bacteriological loops.
Have Questions?
Catalog NumberQuantity
R46012505 Loops/Pk
Catalog number R4601250
Price (USD)
-
Quantity:
5 Loops/Pk

Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.

Bacteroides fragilis ATCC® 25285™ is recommended to be used for culture media and antimicrobial susceptibility quality control testing.

Bacteroides fragilis ATCC® 25285™ is recommended to be used for culture media  and antimicrobial susceptibility quality control testing by CLSI. β-lactamase positive strain.

Technique:

The loop may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000. Alternatively, it can be streaked directly onto appropriate media.

Includes:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

Specifications
ATCC®ATCC™ 25285™
DescriptionBacteroides fragilis ATCC™ 25285™
FormatCulti-Loops
Quantity5 Loops/Pk
Strain DesignationsNCTC 9343, CN 6300, DSM 2151, ONSLOW, VPI 2553, EN-2
Target Organism ClassGram-negative, Rod-shaped Anaerobes
Test TypeQualitative QC
Unit SizeEach