This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Aspergillus flavus ATCC™ 204304™
Thermo Scientific™
Culti-Loops™ Aspergillus flavus ATCC™ 204304™
Perform quality control testing and performance evaluation of microbiological procedures with these ready-to-use bacteriological loops
Have Questions?
Catalog NumberQuantity
R46013125 Loops/Pk.
Catalog number R4601312
Price (USD)
-
Quantity:
5 Loops/Pk.
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Aspergillus flavus ATCC™ 204304™ is recommended to be used for antifungal susceptibility testing quality control. The strain is recommended for antifungal MIC applications QC testing by CLSI and EUCAST.
The loop may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000. Alternatively, it can be streaked directly onto appropriate media.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ATCC®ATCC™ 204304™
DescriptionAspergillus flavus ATCC™ 204304™
FormatCulti-Loops™
Quantity5 Loops/Pk.
Strain DesignationsMCV-C#1
Target Organism ClassFungi
Test TypeQualitative QC
Unit SizeEach