Culti-Loops™ Legionella anisa NCTC 11974™
Culti-Loops™ Legionella anisa NCTC 11974™
Perform quality control testing and performance evaluation of microbiological procedures with these ready-to-use bacteriological loops.
製品番号(カタログ番号)数量
R46013155 Loops/Pk.
製品番号(カタログ番号) R4601315
価格(JPY)
-
数量:
5 Loops/Pk.
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Legionella anisa NCTC 11974™ is recommended as a quality control strain for culture media in water quality testing according to ISO 11731:2016.
The loop can be streaked directly onto appropriate media.
Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
In Vitro 診断にのみご使用ください。
In Vitro 診断にのみご使用ください。
仕様
ATCC®NCTC 11974™
概要Legionella anisa NCTC 11974™
フォーマットCulti-Loops™
数量5 Loops/Pk.
菌株表示ATCC 35292, CCUG 29665, CIP 103870, DSM 17627, WDCM 00106
対象微生物クラスGram-negative Bacilli
テストタイプQualitative QC
Unit SizeEach