This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Klebsiella pneumoniae ATCC™ BAA-2814™
Thermo Scientific™

Culti-Loops™ Klebsiella pneumoniae ATCC™ BAA-2814™

Perform quality control testing and performance evaluation of microbiological procedures with these ready-to-use bacteriological loops.

Have Questions?
Catalog NumberQuantity
R46013165 Loops/Pk.
Catalog number R4601316
Price (USD)
-
Quantity:
5 Loops/Pk.

Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.

Klebsiella pneumoniae ATCC™ BAA-2814™ is recommended to be used for antimicrobial susceptibility testing quality control with disc diffusion and MIC applications.

The Klebsiella pneumoniae ATCC™ BAA-2814™ strain produces Klebsiella pneumoniae carbapenemase KPC-3 and β-lactamase enzymes SHV-11 & TEM-1. The strain is recommended for β-lactam combination agent QC testing, such as Meropenem-vaborbactam and Imipenem-relebactam, by CLSI and EUCAST.

Technique:

The loop can be streaked directly onto appropriate media.

Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.

Includes:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ATCC®ATCC™ BAA-2814™
DescriptionKlebsiella pneumoniae ATCC™ BAA-2814™
FormatCulti-Loops™
Quantity5 Loops/Pk.
Strain DesignationsKP1074 [B21]
Target Organism ClassGram-negative Rods
Test TypeQualitative QC
Unit SizeEach