This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Clostridium perfringens ATCC™ 13124™
Thermo Scientific™
Culti-Loops™ Clostridium perfringens ATCC™ 13124™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46016005 Loops/Pack
Catalog number R4601600
Price (USD)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®13124TM*
DescriptionClostridium perfringens
FormatCulti-Loops
Quantity5 Loops/Pack
Strain DesignationsNCTC 8237 [ATCC 19408, CIP 103 409, CN 1491, NCIB 6125, NCTC 6125, S 107]
Product TypeCulti-Loops Quality Control Organism
SpeciesClostridium species
Target Organism ClassGram-positive Rods
Test TypeQualitative QC
Unit SizeEach