This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Nocardia farcinica ATCC™ 3308™
Thermo Scientific™
Culti-Loops™ Nocardia farcinica ATCC™ 3308™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberProduct Type
R4607156Culti-Loops Quality Control Organism
Catalog number R4607156
Price (USD)
-
Product Type:
Culti-Loops Quality Control Organism
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®3308TM*
DescriptionNocardia farcinica ATCC™ 3308™
FormatCulti-Loops
Packaging Type5 Loops/Package
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassGram-positive Rods
Test TypeQualitative QC
Unit SizeEach