Culti-Loops™ Candida utilis ATCC™ 9950™
Thermo Scientific™
Culti-Loops™ Candida utilis ATCC™ 9950™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Número do catálogoQuantity
R46080125 Loops/Pack
Número do catálogo R4608012
Preço (BRL)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Especificações
ATCC®9950TM*
DescriptionCandida utilis
FormatCulti-Loops
Quantity5 Loops/Pack
Strain DesignationsNRRL Y-900, [CBS 5609, CCRM 20325, DSM 2361, IFO 0988, NBRC 707, NRCC 2721, VTT C-78085]
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassFungi
Test TypeQualitative QC
Unit SizeEach