Culti-Loops™ Fusobacterium necrophorum ATCC™ 25286™
Culti-Loops™ Fusobacterium necrophorum ATCC™ 25286™
Thermo Scientific™
Culti-Loops™ Fusobacterium necrophorum ATCC™ 25286™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46093755 Loops/Pack
Catalog number R4609375
Price (MXN)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®25286™*
DescriptionFusobacterium necrophorum ATCC™ 25286™
FormatCulti-Loops
Quantity5 Loops/Pack
Strain DesignationsVPI 2891 [2358, JCM 3718]
Product TypeFusobacterium species
SpeciesFusobacterium species
Target Organism ClassAnaerobes
Unit SizeEach