This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ API™ Coryne QC Set
Thermo Scientific™
Culti-Loops™ API™ Coryne QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46530634 Loops/Pack
Catalog number R4653063
Price (USD)
-
Quantity:
4 Loops/Pack
Thermo Scientific™ Culti-Loops™ API Coryne Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Cellulosimicrobium cellulans ATCC™ 27402™†
Corynebacterium renale ATCC™ 19412™†
Microbacterium testaceum ATCC™ 15829™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®27402TM*
DescriptionAPI Coryne QC Set/3
FormatCulti-Loops
Quantity4 Loops/Pack
Product TypeCulti-Loops QC Set
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach