This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Sensititre™ Automated Gram Negative QC Set
Thermo Scientific™
Culti-Loops™ Sensititre™ Automated Gram Negative QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46870236 x 1 Loops
Catalog number R4687023
Price (USD)
-
Quantity:
6 x 1 Loops
Thermo Scientific™ Culti-Loops™ Sensititre Automated Gram Negative Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Edwardsiella tarda ATCC™ 15947 ™†
Klebsiella oxytoca ATCC™ 8724™ †
Morganella morganii subsp. morganii ATCC™ 25830™ †
Proteus vulgaris ATCC™ 6896™ †
Pseudomonas aeruginosa ATCC™ 10145 ™†
Shigella sonnei ATCC™ 25931™ †
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®15947TM*
DescriptionSensititre™ Automated Gram Negative QC Set/6
FormatCulti-Loops
Packaging Type6 Loops/Pack
Quantity6 x 1 Loops
Product TypeQC set
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach