This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Sensititre™ YeastOne™ QC Set
Thermo Scientific™
Culti-Loops™ Sensititre™ YeastOne™ QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46870272 x 1 Loop
Catalog number R4687027
Price (USD)
-
Quantity:
2 x 1 Loop
Thermo Scientific™ Culti-Loops™ Sensititre YeastOne Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Candida parapsilosis ATCC™ 22019 ™†
Issatchenkia orientalis ATCC™ 6258 ™†
Ordering Information:
Individually sealed in a foil pouch and packaged two loops (one loop per organism) per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®22019TM*
DescriptionSensititre™ YeastOne QC Set/2
FormatCulti-Loops
Quantity2 x 1 Loop
Product TypeQC set
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach