This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Quanti-Cult Plus™ Aspergillus brasiliensis ATCC™ 16404™
Quanti-Cult Plus™ Aspergillus brasiliensis ATCC™ 16404™
Quanti-Cult Plus™ Aspergillus brasiliensis ATCC™ 16404™
Thermo Scientific™
Quanti-Cult Plus™ Aspergillus brasiliensis ATCC™ 16404™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog NumberQuantity
R4711100100 Tests/Kit
Catalog number R4711100
Price (USD)
-
Quantity:
100 Tests/Kit
Thermo Scientific™ Quanti-Cult Plus™ Aspergillus brasiliensis ATCC™ 16404™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®16404TM*
DescriptionAspergillus brasiliensis ATCC™ 16404™
FormatQuanti-Cult Plus
Packaging Type100 tests (10 vials x 10 tests ea.)
Quantity100 Tests/Kit
Strain DesignationsWLRI 034(120) [CBS 733.88, DSM 1387, DSM 1988, IFO 9455, IMI 149007, NCPF 2275]
Product TypeQuality Control Organism
Target Organism ClassFungi
Test TypeQuantitative QC
Unit SizeEach