Quanti-Cult Plus™ Pseudomonas paraeruginosa ATCC™ 9027™
Quanti-Cult Plus™ Pseudomonas paraeruginosa ATCC™ 9027™
Quanti-Cult Plus™ Pseudomonas paraeruginosa ATCC™ 9027™
Thermo Scientific™
Quanti-Cult Plus™ Pseudomonas paraeruginosa ATCC™ 9027™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog NumberQuantity
R4715210100 Tests/Kit
Catalog number R4715210
Price (CLP)
-
Quantity:
100 Tests/Kit
Thermo Scientific™ Quanti-Cult Plus™ Pseudomonas paraeruginosa ATCC™ 9027™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • Ready for rehydration
  • For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®9027™*
DescriptionPseudomonas paraeruginosa ATCC™ 9027™
FormatQuanti-Cult Plus
Quantity100 Tests/Kit
Product TypeQuality Control Organism
Target Organism ClassGram-negative Rods
Test TypeQuantitative QC
Unit SizeEach