This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Quanti-Cult Plus™ Burkholderia cepacia ATCC™ 25416™
Quanti-Cult Plus™ Burkholderia cepacia ATCC™ 25416™
Quanti-Cult Plus™ Burkholderia cepacia ATCC™ 25416™
Thermo Scientific™
Quanti-Cult Plus™ Burkholderia cepacia ATCC™ 25416™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog NumberQuantity
R4715220100 Tests/Kit
Catalog number R4715220
Price (USD)
-
Quantity:
100 Tests/Kit
Thermo Scientific™ Quanti-Cult Plus™ Burkholderia cepacia ATCC™ 25416™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • Ready for rehydration
  • For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®25416TM*
DescriptionBurkholderia cepacia ATCC™ 25416™
FormatQuanti-Cult Plus
Packaging Type100 Test/Kit
Quantity100 Tests/Kit
Strain DesignationsUCB 717 [717-ICPB 25, Ballard 717, CCUG 12691, CCUG 13226, CFBP 2227, CIP 80.24, DSM 7288, HAMBI 1976, ICMP 5796, IFO 14074, JCM 5964, LMG 1222, NCCB 76047, NCPPB 2993, NCTC 10743, NRRL B-14810]
Product TypeQuality Control Organism
Target Organism ClassGram-negative Rods
Test TypeQuantitative QC
Unit SizeEach