Quanti-Cult Plus™ Salmonella sp. serovar Abony NCTC 6017
Quanti-Cult Plus™ Salmonella sp. serovar Abony NCTC 6017
Quanti-Cult Plus™ Salmonella sp. serovar Abony NCTC 6017
Thermo Scientific™
Quanti-Cult Plus™ Salmonella sp. serovar Abony NCTC 6017
Supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Have Questions?
Número do catálogoQuantity
R4716007100 Tests/Kit
Número do catálogo R4716007
Preço (BRL)
-
Quantity:
100 Tests/Kit
Thermo Scientific™ Quanti-Cult Plus is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • Ready for rehydration
  • For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Especificações
ATCC®NCTC 6017
DescriptionSalmonella sp. serovar Abony NCTC 6017
FormatQuanti-Cult Plus
Packaging Type100 Test/Kit
Quantity100 Tests/Kit
Strain DesignationsCCM 4518 [NBRC 100797, NCTC 6017, CIP 80.39]
Product TypeQuality Control Organism
Target Organism ClassGram-negative Rods
Test TypeQuantitative QC
Unit SizeEach