This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Quanti-Cult™ Staphylococcus aureus subsp. aureus ATCC™ 25923™
Thermo Scientific™
Quanti-Cult™ Staphylococcus aureus subsp. aureus ATCC™ 25923™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog NumberQuantity
R473701010 Tests/Kit
Catalog number R4737010
Price (USD)
-
Quantity:
10 Tests/Kit
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media that can be stored at room-temperature to free up incubator space.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum
Ordering Alerts:
Yiels 1 Test/reconstituted vial, 10 Tests/kit
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®25923TM*
DescriptionStaphylococcus aureus subsp. aureus ATCC™ 25923™
FormatQuanti-Cult
Packaging Type10 Test/Kit
Quantity10 Tests/Kit
Strain DesignationsSeattle 1944
Product TypeBactidisk
Target Organism ClassCocci
Test TypeQuantitative QC
Unit SizeEach