This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
WaterBugs™ Enterobacter cloacae subsp. cloacae ATCC™ 13047™
Thermo Scientific™
WaterBugs™ Enterobacter cloacae subsp. cloacae ATCC™ 13047™
Use in water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult™ format.
Have Questions?
Catalog NumberQuantity
R475709010 Tests/Kit
Catalog number R4757090
Price (USD)
-
Quantity:
10 Tests/Kit
Thermo Scientific™ WaterBugs™ Enterobacter cloacae subsp. cloacae ATCC™ 13047™ is used in water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
  • Deliver <50 CFUs in a single inoculum to mimic water samples
  • One test/reconstituted vial
Ordering Alerts:
Supplied with 10 preserved organism vials; 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®13047TM*, 13047™*
DescriptionEnterobacter cloacae subsp. cloacae ATCC™ 13047™
FormatQuanti-Cult
Packaging Type10 Test/Kit
Quantity10 Tests/Kit
Product TypeQuality Control Organism
Target Organism ClassGram-negative Rods
Test TypeQuantitative QC
Unit SizeEach