Product Image
Thermo Scientific™
Geobacillus stearothermophilus ATCC™ 7953™
Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoATCC®Description
R48510017953TM*Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population
R48510027953TM*Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 106spore population
Número do catálogo R4851001
Preço (BRL)
-
ATCC®:
7953TM*
Description:
Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population
Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.
  • 10 -6spore population
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Especificações
ATCC®7953TM*
DescriptionSpore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population
FormatSpore Strips
Product TypeQuality Control Organism
Target Organism ClassGram-positive Rods
Test TypeQualitative QC
Unit SizeEach