Disposable Calibrated Inoculating Loops
Actual product may vary
Thermo Scientific™
Disposable Calibrated Inoculating Loops
Select, inoculate or transfer microbiological sample volume on media plates with these calibrated inoculating loops.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolumeProduct TypeColor
R5014201 μLInoculating LoopBlue
R501400No LoopInoculating NeedleWhite
R50141010 μLInoculating LoopYellow
Catalog number R501420
Price (USD)
-
Volume:
1 μL
Product Type:
Inoculating Loop
Color:
Blue
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Disposable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
Eliminate the need for flaming or sterilization with these sterile, disposable loops.
For Laboratory Use
For Laboratory Use
Specifications
CalibratedYes
ColorBlue
DisposableSingle-use
Handle TypePlastic
Quantity30 bags, 32 units/bag
MaterialPlastic (Handle)
Product TypeInoculating Loop
SterilitySterile
Volume1 μL
Unit SizeEach