Reusable Calibrated Inoculating Loops
Reusable Calibrated Inoculating Loops
Thermo Scientific™
Reusable Calibrated Inoculating Loops
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeHandle TypeSizeQuantity
R503060Inoculating NeedlePurple Insulated HandleEach
R503152Inoculating LoopWhite Insulated Handle2 mm5/Pk.
R503154Inoculating LoopGreen Insulated Handle3 mmEach
R503155Inoculating LoopYellow Insulated Handle5 mmEach
R503620Inoculating LoopWithout Handle1/1000 mLEach
R507010Inoculating Loop Calibration GaugeNot Applicable0.01 mLEach
R501966Insulated Brass HolderBrass Loop Holder6 in.Each
R501968Insulated Brass HolderBrass Loop Holder8 in.Each
R503070Teasing NeedleYellow Insulated HandleEach
R503080Teasing NeedleOrange Insulated HandleEach
R503090Inoculating TB SpadeBlue Insulated HandleEach
R503151Inoculating LoopWhite Insulated Handle2 mmEach
R503153Inoculating LoopGreen Insulated Handle3 mmEach
R503156Inoculating LoopYellow Insulated Handle5 mm5/Pk.
R503513Inoculating LoopWithout handle3 mm25/Pk.
R503515Inoculating LoopWithout handle5 mm25/Pk.
R503630Inoculating LoopWithout Handle1/100 mLEach
R503730Inoculating LoopBlue Insulated Handle1/1000 mLEach
R503740Inoculating LoopOrange Insulated Handle1/100 mLEach
R507020Inoculating Loop Calibration GaugeNot Applicable0.001 mLEach
Catalog number R503060
Price (USD)
-
Request A Quote
Product Type:
Inoculating Needle
Handle Type:
Purple Insulated Handle
Quantity:
Each
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
Reusable loops feature options with and without lightweight aluminum handles, constructed of high-quality nichrome wire which cools rapidly after sterilization, and the added benefit of additional insulation for hand protection.
For Laboratory Use
For Laboratory Use
Specifications
ColorPurple
Handle TypePurple Insulated Handle
QuantityEach
MaterialNichrome
Product TypeInoculating Needle
VolumeNo Loop
Unit SizeEach