Product Image
COPAN WASP™DT Instrument Accessories and Software, Inoculation Broth Module
Build a more efficient microbiology laboratory and automate all aspects of specimen processing with COPAN WASP™ Instrument Accessories and Software.
Have Questions?
Catalog number R5086011
Price (USD)
-
Build a more efficient microbiology laboratory and automate all aspects of specimen processing with COPAN WASP™ Instrument Accessories and Software. Structure the modular system according to the unique needs of your laboratory, and choose from a variety of instrument accessories, from enrichment broth inoculation to an automatic loop and tool changing station, to drive productivity from start to finish as well as maximize current and future investments.
The WASP System works seamlessly with Thermo Scientific™ Remel™ prepared media products which are optimized for automation.
Addressing all aspects of microbiology specimen processing with standardized automation for consistent quality, the WASP System's modular platform ensures optimized lab productivity.
  • Up to 378 plate capacity increases walk-away time—for high-throughput and faster returns on investment, use Thermo Scientific Remel bi-plates
  • Multi-media carousel processes various culture setups using up to nine different types of media
  • Dynamic pick and place robotics mimic workflow routinely performed by lab technicians and prevents cross contamination
  • Continuous and random loading eliminates the need for batching
  • Specimen verification system ensures quality
  • Smart Scan Technology reduces errors and streamlines traceability 
  • Proprietary gel-preservation ensures faster recovery with Thermo Scientific™ Culti-Loops™ Quality Control Organisms
Get the most out of your automated specimen processing system with Thermo Scientific Remel prepared media products. Manufactured in state-of-the-art facilities and supported by a responsive team of microbiology experts, Remel prepared media is the preferred choice of laboratories around the world. With more than 1,700 formats and formulations designed to deliver unmatched recovery and reproducibility, you can rest assured knowing that your automated specimen processing is working with products of the highest quality and consistency.
COPAN WASP instrumentation and accessories are available through Thermo Fisher Scientific in the U.S.A. only. Please contact your local sales representative for more information.
ESwab and WASP are trademarks or registered trademarks of COPAN S.P.A. All other trademarks are property of Thermo Fisher Scientific and its subsidiaries.
Specifications
DescriptionWASP™DT Inoculation Broth Module
QuantityEa.
Product TypeInoculation Broth Module
Unit SizeEach