PathoDx Strep Grouping Universal Kit
Thermo Scientific™
PathoDx Strep Grouping Universal Kit
Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R62150120 Tests/Kit
R6207660 Tests/Kit
Catalog number R62150
Price (MXN)
-
Quantity:
120 Tests/Kit
Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit. This kit is a set of components to be used with selected, separately purchased PathoDxStrep Grouping Reagents designed to identify beta-haemolytic streptococci of Lancefield Groups A, B, C, F or G from primary culture plates.This accessory kit contains: Extraction reagent 1; Extraction reagent 2; Extraction reagent 3; Positive control; Reaction cards; Stirrers.
The PathoDx procedure utilizes a latex agglutination method in conjunction with a nitrous acid extraction procedure.1,2 The IgG coupled to the latex is highly specific for a given streptococcal group antigen. This method offers significant advantages over other streptococcal grouping procedures in terms of rapidity, simplicity, and convenience.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
1.El Kholy, A., et al. 1974. Appl. Microbiol. 28:836-839.
2.Hryniewicz, W., et al. 1976. J. Clin. Microbiol. 4:28-31.
Specifications
Control SetsPositive
DescriptionStrep Universal Grouping Kit
IncludesControl antigens ABC and FG; extraction reagents 1, 2, 3; stirrers; disposable slides
Organism GroupLancefield A, B, C, F or G
Quantity120 Tests/Kit
TypeStreptococcus Test Kit
Detectable Analytesbeta-haemolytic Streptococci (Groups A, B, C, F, G)
Unit SizeEach