See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Cell Culture Utensils
▸
Inoculating Loops and Needles
▸
Remel™ Microstreaker Loop
Thermo Scientific™
Remel™ Microstreaker Loop
Thermo Scientific™ Remel Microstreaker is a twisted nichrome loop with handle.
Have Questions?
Change view
Catalog Number
Size
Quantity
Calibrated
R641851
1/1000
1 Ea.
Yes
R641800
Large
5/Pk.
No
R641840
Small
5/Pk.
No
R641820
Large
5/Pk.
R641871
1/100
1 Ea.
Yes
5 Options
Catalog number
R641851
Price (USD)
-
Request A Quote
Size:
1/1000
Quantity:
1 Ea.
Calibrated:
Yes
Thermo Scientific™ Remel Microstreaker is a twisted nichrome loop with handle.
Specifications
Calibrated
Yes
Quantity
1 Ea.
Material
Nichrome
Size
1/1000
Volume
0.001 μL