Product Image
Thermo Scientific™

Denka Seiken™ Agglutinating Sera Vibrio parahaemolyticus K

Denka Seiken Agglutinating Sera, Vibrio parahaemolyticus K are packaged 2mL per vial.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
R671405Vibrio parahaemolyticus K VII, types K48, 49, 50, 51, 52, 53, 54
R671573Vibrio parahaemolyticus K18
R671931Vibrio parahaemolyticus K56
R671429Vibrio parahaemolyticus K IX, types K63, 64, 65, 66, 67, 68, 69
R671474Vibrio parahaemolyticus K6
R671044Vibrio parahaemolyticus K68, types K1, 3, 4, 5, 6, 7, 8
R674535Vibrio parahaemolyticus K12
R671436Vibrio parahaemolyticus K1
R671351Vibrio parahaemolyticus K II, types K9, 10, 11, 12, 13, 15, 17
R671412Vibrio parahaemolyticus K VIII, types K55, 56, 57, 58, 59, 60, 61
R671900Vibrio parahaemolyticus K53
R671719Vibrio parahaemolyticus K33
R671924Vibrio parahaemolyticus K55
Catalog number R671405
Price (USD)
-
Type:
Vibrio parahaemolyticus K VII, types K48, 49, 50, 51, 52, 53, 54

Denka Seiken Agglutinating Sera, Vibrio parahaemolyticus K are packaged 2mL per vial.

Specifications
FormatVial
Quantity2 mL/Vial
TypeVibrio parahaemolyticus K VII, types K48, 49, 50, 51, 52, 53, 54
CE MarkerYes
Unit SizeEach