Product Image
Thermo Scientific™

Denka Seiken™ Agglutinating Sera Salmonella Antisera Sets

Denka Seiken Agglutinating Sera are packaged 2mL per vial.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
R679916Salmonella Antisera Set 7
R679886Salmonella Antisera Set 4
R679893Salmonella Antisera Set 5
R679855Salmonella Antisera Set 1
R679909Salmonella Antisera Set 6
R679862Salmonella Antisera Set 2
R679879Salmonella Antisera Set 3
Catalog number R679916
Price (USD)
-
Description:
Salmonella Antisera Set 7

Denka Seiken Agglutinating Sera are packaged 2mL per vial.

  • Contains O Groups O2, O9; H groups a, d, and Vi
Specifications
DescriptionSalmonella Antisera Set 7
IncludesO Groups O2, O9; H groups a, d, and Vi
Organism GroupGroup O2, O9
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella
TypeAgglutinating Sera
Unit SizeEach