Product Image
Thermo Scientific™
AnaeroPack™ Rectangular Jar Body
Mitsubishi™ AnaeroPack Rectangular Jar Body is a replacement part for anaerobic jar.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
R6850777.0 L
R6850070.4 L
R6850272.5 L
Catalog number R685077
Price (USD)
-
Volume (Metric):
7.0 L
Mitsubishi™ AnaeroPack Rectangular Jar Body is a replacement part for anaerobic jar.
Specifications
QuantityEach
Volume (Metric)7.0 L
FormatRectangular Jar
Product LineThermo Scientific Mitsubishi
TypeAnaeroPack Rectangular Jar Body
Unit SizeEach