Remel™ BactiSwab™ Gel Collection and Transport Swabs: Single Swab Amies Charcoal
Thermo Scientific™
Remel™ BactiSwab™ Gel Collection and Transport Swabs: Single Swab Amies Charcoal
Thermo Scientific™ Remel BactiSwab Gel Collection and Transport Swabs provide excellent recovery for a wide range of bacteria, including anaerobes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging
R723040500/Pk.
R72304550/Pk.
Catalog number R723040
Price (CLP)
-
Packaging:
500/Pk.
Thermo Scientific™ Remel BactiSwab Gel Collection and Transport Swabs provide excellent recovery for a wide range of bacteria, including anaerobes.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Recommended for recovery of Neisseria gonorrhoeae
  • Single plastic shaft
  • Black cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
Specifications
Closure ColorBlack
DescriptionAmies Charcoal
Packaging500/Pk.
Quantity1 Tube/Envelope
Shaft MaterialPlastic
FormatTube
Media TypeAmies Charcoal
Unit SizeEach