StemPro® Rat Alk Phos Expressing Mesenchymal Stem Cells - FAQs
StemPro® Rat Alk Phos Expressing Mesenchymal Stem Cells - FAQs
View additional product information for StemPro® Rat Alk Phos Expressing Mesenchymal Stem Cells - FAQs (R7789120)
3 product FAQs found
What is the strain of rat used to isolate StemPro Rat Alk Phos-expressing MSCs?
StemPro Rat Alk Phos-expressing MSCs are isolated from the bone marrow of pooled young transgenic Fisher 344 rats.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do all StemPro Alk Phos-expressing Rat MSCs adhere when they are thawed?
A small percentage of cells might not adhere when the cells are first thawed. However if the adherent cells are expanded, the expanded cells will be adherent in future passages.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
For how many passages can the StemPro Alk Phos-expressing Rat MSCs be cultured?
We have tested the cells for expansion up to 3 passages.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.