FreeStyle™ CHO-S Cells are part of the FreeStyle™ MAX Expression System, which is a breakthrough technology for rapid and high-yieldRead more
Have Questions?
Catalog Number
Quantity
R80007
1 mL
Catalog number R80007
Price (USD)
1,160.00
Each
Quantity:
1 mL
Price (USD)
1,160.00
Each
FreeStyle™ CHO-S Cells are part of the FreeStyle™ MAX Expression System, which is a breakthrough technology for rapid and high-yield mammalian protein production. This cell line is adapted to high density, serum-freesuspension culture in FreeStyle™ CHO Expression Medium and iscapable of producing high levels of recombinant protein.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineFreeStyle™ CHO-S Cells
For Use With (Application)cand Transfection
Product LineFreestyle
Product TypeCHO-S Cell
Quantity1 mL
Starting Material Amount1 x 107
System TypeFreeStyle™
Cell TypeCHO-S
SpeciesHamster
Transfection TechniqueLipid-based Transfection
Unit SizeEach
Contents & Storage
1 vial of 1 x 107 cells. Store in liquid nitrogen.
Frequently asked questions (FAQs)
How do FreeStyle 293-F cells compare to FreeStyle CHO-S cells in terms of antibody and protein yield?
When comparing FreeStyle 293-F cells with FreeStyle CHO-S cells for average range of antibody and protein yield, it is definitely expected that antibody yield will be higher in FreeStyle 293-F cells than in FreeStyle CHO-S cells, when generated by transient transfection. On the other hand, antibody yields in stable CHO cells can be much higher. For non-antibody proteins, it is impossible to predict which of the two cell lines may provide higher yields.
How long can I keep culturing FreeStyle cells after transfection?
We typically keep the culture going for 7-10 days posttransfection. After that, protein expression may drop.